| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700007756.mm.1 |
NM_153057.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007756.mm.1 |
NM_153057.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007756.mm.1 |
TC0700000870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007757.mm.1 |
NM_153057.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007757.mm.1 |
NM_153057.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007757.mm.1 |
TC0700000870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007758.mm.1 |
NM_153057.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007758.mm.1 |
NM_153057.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007758.mm.1 |
TC0700000870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007759.mm.1 |
NM_153057.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007759.mm.1 |
NM_153057.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007759.mm.1 |
TC0700000870.mm.1 |
MTA TC |