Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153057.4 | NP_694697.3 | 22 - 23 | 2756 | 71 | |
| GenBank mRNA | AK030584.1 | - | 22 - 23 | 2723 | 71 | |
| AK080754.1 | - | 22 - 23 | 2619 | 71 | ||
| AK082495.1 | - | 22 - 23 | 2635 | 71 | ||
| AK082963.1 | - | 22 - 23 | 2615 | 71 | ||
| AK142338.1 | - | 22 - 23 | 2726 | 71 | ||
| AK150107.1 | - | 22 - 23 | 2711 | 71 | ||
| AK165643.1 | - | 22 - 23 | 2619 | 71 | ||
| AK166764.1 | - | 7 - 8 | 766 | 71 | ||
| BC022138.1 | - | 21 - 22 | 2472 | 71 | ||
| BC025133.1 | - | 5 - 6 | 635 | 71 | ||
| BC033923.1 | - | 11 - 12 | 1340 | 5099899 | 71 | |
| BC072630.1 | - | 22 - 23 | 2702 | 6833516 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map