| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100008284.mm.1 |
NM_153081.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008284.mm.1 |
NM_153081.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008284.mm.1 |
TC1100000943.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008285.mm.1 |
NM_153081.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008285.mm.1 |
NM_153081.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008285.mm.1 |
TC1100000943.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008279.mm.1 |
NM_153081.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008279.mm.1 |
NM_153081.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008279.mm.1 |
TC1100000943.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008273.mm.1 |
NM_153081.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008273.mm.1 |
NM_153081.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008273.mm.1 |
TC1100000943.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008272.mm.1 |
NM_153081.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008272.mm.1 |
NM_153081.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008272.mm.1 |
TC1100000943.mm.1 |
MTA TC |