Assay Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153081.3 | NP_694721.2 | 3 - 4 | 709 | 101 | |
| GenBank mRNA | AB041591.1 | - | 3 - 4 | 502 | 101 | |
| BC130255.1 | - | 2 - 3 | 212 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899473 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061338 | BE990575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061338 | NM_153081.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2061338 | AB041591.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804246 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124940 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061338 | BE990575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061338 | NM_153081.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2061338 | AB041591.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27899472 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124940 | - | - |
| Whole Mouse Genome G4122A | A_51_P489720 | BE990575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489720 | NM_153081.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489720 | AB041591.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139847 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124940 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09641177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701637 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701632 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701649 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09701639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09641182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27899471 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377798 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427107_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377790 | - | - |
| Mouse Genome 430 2.0 Array | 1427107_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427107_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216867_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216867_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100008293.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008293.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008293.mm.1 | TC1100000943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008295.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008295.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008295.mm.1 | TC1100000943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008296.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008296.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008296.mm.1 | TC1100000943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008298.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008298.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008298.mm.1 | TC1100000943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008292.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008292.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008292.mm.1 | TC1100000943.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100008294.mm.1 | NM_153081.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008294.mm.1 | NM_153081.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008294.mm.1 | TC1100000943.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427107_PM_at | - | - |
| Mouse Expression Set 430 | 1427107_at | - | - |
| Murine Genome U74v2 Set | 139487_PM_at | - | - |
| Murine Genome U74v2 Set | 139487_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427107_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427107_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427107_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427107_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 491270 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6380369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721413 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721413 | - | - |
| mouse-6 v1.1 (Illumina) | scl41341.3_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41341.3_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2721413 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721413 | - | - |

Slc16a11
Genomic Map