Assay Details
| Gene Symbol: Abca8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153145.4 | NP_694785.3 | 17 - 18 | 2344 | 74 | |
| GenBank mRNA | AB255881.1 | - | 16 - 17 | 2263 | 74 | |
| AB255882.1 | - | 17 - 18 | 2351 | 74 | ||
| AF498360.1 | - | 16 - 17 | 2272 | 74 | ||
| AY732492.1 | - | 16 - 17 | 2272 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca8a
Genomic Map