Assay Details
| Gene Symbol: P2rx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164833.1 | NP_001158305.1 | 4 - 5 | 539 | 66 | |
| NM_001164834.1 | NP_001158306.1 | 4 - 5 | 539 | 66 | ||
| NM_153400.4 | NP_700449.2 | 4 - 5 | 539 | 66 | ||
| GenBank mRNA | AB094663.1 | - | 4 - 5 | 459 | 66 | |
| AB094664.1 | - | 4 - 5 | 459 | 66 | ||
| AK141196.1 | - | 4 - 5 | 498 | 66 | ||
| AK158334.1 | - | 4 - 5 | 517 | 66 | ||
| AY044240.2 | - | 4 - 5 | 715 | 66 | ||
| BC106963.1 | - | 4 - 5 | 346 | 66 | ||
| BC106964.1 | - | 4 - 5 | 346 | 40039097 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx2
Genomic Map