Assay Details
| Gene Symbol: P2rx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164833.1 | NP_001158305.1 | 7 - 8 | 859 | 62 | |
| NM_001164834.1 | NP_001158306.1 | 7 - 8 | 859 | 62 | ||
| NM_153400.4 | NP_700449.2 | 7 - 8 | 859 | 62 | ||
| GenBank mRNA | AB094663.1 | - | 7 - 8 | 779 | 62 | |
| AB094664.1 | - | 7 - 8 | 779 | 62 | ||
| AK158334.1 | - | 7 - 8 | 837 | 62 | ||
| AY044240.2 | - | 7 - 8 | 1035 | 62 | ||
| BC106963.1 | - | 6 - 7 | 750 | 62 | ||
| BC106964.1 | - | 7 - 8 | 666 | 40039097 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
transport
ion transport
chemical synaptic transmission
neuromuscular synaptic transmission
neuromuscular junction development
sensory perception of sound
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
behavioral response to pain
skeletal muscle fiber development
sensory perception of taste
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
copper ion binding
ATP binding
drug binding
zinc ion binding
ligand-gated ion channel activity
nickel cation binding
phosphatidylinositol binding
identical protein binding
mercury ion binding
cadmium ion binding
cobalt ion binding
response to hypoxia
detection of hypoxic conditions in blood by carotid body chemoreceptor signaling
transport
ion transport
chemical synaptic transmission
neuromuscular synaptic transmission
neuromuscular junction development
sensory perception of sound
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
behavioral response to pain
skeletal muscle fiber development
sensory perception of taste
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ion channel activity
copper ion binding
ATP binding
drug binding
zinc ion binding
ligand-gated ion channel activity
nickel cation binding
phosphatidylinositol binding
identical protein binding
mercury ion binding
cadmium ion binding
cobalt ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979132 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182975 | AK158334.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182975 | AY044240.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182975 | AB094663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182975 | AB094664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182975 | AK141196.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038728 | BC106964.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038728 | AK158334.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038728 | AY044240.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038728 | BC106963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038728 | AB094663.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971227 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144796 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182975 | AK158334.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182975 | AY044240.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182975 | AB094663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182975 | AB094664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182975 | AK141196.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038728 | BC106964.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038728 | AK158334.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038728 | AY044240.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038728 | BC106963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038728 | AB094663.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498526 | BC106964.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498526 | AK158334.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498526 | AY044240.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498526 | BC106963.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P498526 | AB094663.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23979151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144796 | - | - |
| Whole Mouse Genome G4122A | A_51_P498526 | BC106964.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498526 | AK158334.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498526 | AY044240.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498526 | BC106963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P498526 | AB094663.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128382 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355035 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04355069 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979134 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979133 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23979132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23979122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04764425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04131442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04860049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04554268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04860054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04860055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04860052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04355011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979131 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532356 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532357 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532365 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435212_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532346 | - | - |
| Mouse Genome 430 2.0 Array | 1435212_at | - | - |
| Mouse Genome 430 2.0 Array | 1435212_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231602_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231602_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_001164833.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_001164833.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_001164834.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_001164834.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_153400.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | NM_153400.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500026433.mm.1 | TC0500002988.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435212_PM_at | - | - |
| Mouse Expression Set 430 | 1435212_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435212_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435212_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3840064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2735318 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2735318 | - | - |
| mouse-6 v1.1 (Illumina) | scl26228.11_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26228.11_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2735318 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2735318 | - | - |
Citations (1)
Comparison of P2X and TRPV1 receptors in ganglia or primary culture of trigeminal neurons and their modulation by NGF or serotonin.
- Authors
- Simonetti M, Fabbro A, D'Arco M, Zweyer M, Nistri A, Giniatullin R, Fabbretti E
- Journal
- Mol Pain - Page No 11 (Sun Jan 01 00:00:00 UTC 2006)
- Volume (Year)
- 2 (2006)
- Product Usage
- -

P2rx2
Genomic Map