Assay Details
| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153403.2 | NP_700452.2 | 3 - 4 | 331 | 98 | |
| GenBank mRNA | AB081471.1 | - | 3 - 4 | 331 | 98 | |
| AK080954.1 | - | 1 - 2 | 99 | 98 | ||
| BC129914.1 | - | 3 - 4 | 331 | 40111808 | 98 | |
| BC129915.1 | - | 3 - 4 | 327 | 98 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK080954.1 transcripts.
Set Membership: |
|

Ago1
Genomic Map