Assay Details
| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153520.1 | NP_705740.1 | 5 - 6 | 334 | 104 | |
| GenBank mRNA | AK078150.1 | - | 527 | 104 | ||
| AK140031.1 | - | 335 | 104 | |||
| BC060980.1 | - | 5 - 6 | 293 | 6533846 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163903 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25320 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136432 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125011 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135754 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P262563 | AK078150.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32112848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125011 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135754 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136432 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09172578 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03597255 | - | - |
| Whole Mouse Genome G4122A | A_51_P262563 | AK078150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | AK078150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | AK140031.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | BC060980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P624415 | NM_153520.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110216 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125011 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136432 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14339358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32112792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14303604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14475008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14488938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14378550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14488936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14319721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14167283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14285822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14340798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14448757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14503391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14340812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32112844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10467532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467537 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467531 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435854_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447487_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10467529 | - | - |
| Mouse Genome 430 2.0 Array | 1447487_at | - | - |
| Mouse Genome 430 2.0 Array | 1435854_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447487_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435854_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226115_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226115_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900013132.mm.1 | NM_153520.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013132.mm.1 | NM_153520.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900013132.mm.1 | TC1900001503.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1447487_at | - | - |
| Mouse Expression Set 430 | 1447487_PM_at | - | - |
| Mouse Expression Set 430 | 1435854_PM_at | - | - |
| Mouse Expression Set 430 | 1435854_at | - | - |
| Murine Genome U74v2 Set | 166890_PM_at | - | - |
| Murine Genome U74v2 Set | 135230_PM_at | - | - |
| Murine Genome U74v2 Set | 135230_at | - | - |
| Murine Genome U74v2 Set | 166890_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435854_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447487_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447487_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435854_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3840528 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3840528 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840528 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230025 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230025 | - | - |
| mouse-6 v1.1 (Illumina) | scl0226115.1_174-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840528 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0226115.1_174-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840528 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2824889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2824889 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0226115.1_174-S | - | - |

Opalin
Genomic Map