Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 12 - 13 | 1760 | 97 | |
| GenBank mRNA | AK128970.1 | - | 12 - 13 | 1775 | 97 | |
| BC034070.1 | - | 2 - 3 | 360 | 4217923 | 97 | |
| BC040815.1 | - | 2 - 3 | 360 | 4217923 | 97 | |
| BC067035.1 | - | 12 - 13 | 1760 | 30532658 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map