Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 22 - 23 | 3006 | 79 | |
| GenBank mRNA | AK088754.1 | - | 7 - 8 | 879 | 79 | |
| AK128970.1 | - | 22 - 23 | 3021 | 79 | ||
| AK129086.1 | - | 2658 | 79 | |||
| AK163968.1 | - | 5 - 6 | 630 | 79 | ||
| BC031364.1 | - | 8 - 9 | 1020 | 79 | ||
| BC034070.1 | - | 12 - 13 | 1606 | 4217923 | 79 | |
| BC040815.1 | - | 12 - 13 | 1606 | 4217923 | 79 | |
| BC067035.1 | - | 22 - 23 | 3006 | 30532658 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map