Assay Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153548.2 | NP_705776.2 | 24 - 25 | 3191 | 58 | |
| GenBank mRNA | AK088754.1 | - | 9 - 10 | 1064 | 58 | |
| AK128970.1 | - | 24 - 25 | 3206 | 58 | ||
| AK129086.1 | - | 2843 | 58 | |||
| AK163968.1 | - | 7 - 8 | 815 | 58 | ||
| BC031364.1 | - | 10 - 11 | 1205 | 58 | ||
| BC034070.1 | - | 14 - 15 | 1791 | 4217923 | 58 | |
| BC040815.1 | - | 14 - 15 | 1791 | 4217923 | 58 | |
| BC067035.1 | - | 24 - 25 | 3191 | 30532658 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

E430025E21Rik
Genomic Map