Assay Details
| Gene Symbol: Nipa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153578.2 | NP_705806.1 | 2 - 3 | 211 | 102 | |
| GenBank mRNA | AK046619.1 | - | 2 - 3 | 226 | 102 | |
| AK166325.1 | - | 189 | 102 | |||
| AY098645.1 | - | 2 - 3 | 211 | 102 | ||
| BC055828.1 | - | 2 - 3 | 211 | 4192279 | 102 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK166325.1 transcripts.
Set Membership: |
|

Nipa1
Genomic Map