Assay Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009481.2 | NP_033507.2 | 23 - 24 | 3957 | 83 | |
| GenBank mRNA | DQ086491.1 | - | 23 - 24 | 3918 | 83 | |
| U67874.1 | - | 23 - 24 | 3918 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp9x
Genomic Map