Assay Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009481.2 | NP_033507.2 | 35 - 36 | 6480 | 151 | |
| GenBank mRNA | AK034941.1 | - | 1 - 2 | 541 | 151 | |
| BY716634.1 | - | 1 - 2 | 583 | 151 | ||
| DQ086491.1 | - | 35 - 36 | 6441 | 151 | ||
| U67874.1 | - | 35 - 36 | 6441 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp9x
Genomic Map