Assay Details
| Gene Symbol: Pcgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163307.1 | NP_001156779.1 | 6 - 7 | 604 | 66 | |
| NM_001163308.1 | NP_001156780.1 | 5 - 6 | 466 | 66 | ||
| NM_009545.2 | NP_033571.1 | 8 - 9 | 871 | 66 | ||
| GenBank mRNA | AF483502.1 | - | 4 - 5 | 312 | 66 | |
| AF483503.1 | - | 4 - 5 | 312 | 66 | ||
| AK142837.1 | - | 5 - 6 | 379 | 66 | ||
| BC016419.1 | - | 6 - 7 | 558 | 4458285 | 66 | |
| BI991588.1 | - | 5 - 6 | 532 | 66 | ||
| D90085.1 | - | 8 - 9 | 719 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf2
Genomic Map