Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 14 - 15 | 6018 | 74 | |
| GenBank mRNA | AF031572.1 | - | 14 - 15 | 6018 | 74 | |
| AK220170.1 | - | 13 - 14 | 1912 | 74 | ||
More Information
Set Membership: |
|

Celsr1
Genomic Map