Assay Details
| Gene Symbol: Celsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009886.2 | NP_034016.2 | 15 - 16 | 6199 | 92 | |
| GenBank mRNA | AF031572.1 | - | 15 - 16 | 6199 | 92 | |
| AK220170.1 | - | 14 - 15 | 2093 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Celsr1
Genomic Map