Assay Details
| Gene Symbol: Mxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016662.4 | NP_057871.2 | 3 - 4 | 269 | 76 | |
| GenBank mRNA | AK138742.1 | - | 3 - 4 | 270 | 76 | |
| BC051970.1 | - | 3 - 4 | 272 | 5714980 | 76 | |
| U32394.1 | - | 3 - 4 | 206 | 76 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054386 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054387 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054388 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054389 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903658 | BC111822.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903658 | NM_053219.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175419 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176486 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315530 | AK086190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315530 | AK138742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315530 | BC051970.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315530 | U32394.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P315530 | NM_016662.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29054401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07882526 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176486 | - | - |
| Whole Mouse Genome G4122A | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P190647 | AK014516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315530 | AK086190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315530 | AK138742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315530 | BC051970.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315530 | U32394.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P315530 | NM_016662.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118708 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10821707 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10610122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10610123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10610120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10821703 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10610127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054388 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054389 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054386 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054387 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29054390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29054331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10900329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10723420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11057954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10777838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10777836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11109886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10821706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11109885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10438504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10600441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10531359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11054964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11057943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29054377 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10409426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10409428 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422970_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438847_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448878_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10409424 | - | - |
| Mouse Genome 430 2.0 Array | 1422970_at | - | - |
| Mouse Genome 430 2.0 Array | 1422970_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438847_at | - | - |
| Mouse Genome 430 2.0 Array | 1438847_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448878_at | - | - |
| Mouse Genome 430 2.0 Array | 1448878_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17121_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17121_PM_at | - | - |
| Mouse Expression Set 430 | 1438847_PM_at | - | - |
| Mouse Expression Set 430 | 1438847_at | - | - |
| Mouse Expression Set 430 | 1422970_PM_at | - | - |
| Mouse Expression Set 430 | 1422970_at | - | - |
| Mouse Expression Set 430 | 1448878_at | - | - |
| Mouse Expression Set 430 | 1448878_PM_at | - | - |
| Murine Genome U74v2 Set | 103237_at | - | - |
| Murine Genome U74v2 Set | 103237_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438847_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448878_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422970_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422970_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448878_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438847_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448878_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448878_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422970_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422970_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438847_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438847_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 918397 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6220014 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642571 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642571 | - | - |
| mouse-6 v1.1 (Illumina) | scl43929.4.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43929.4.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642571 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642571 | - | - |

Mxd3
Genomic Map