Assay Details
| Gene Symbol: Cdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026014.3 | NP_080290.3 | 7 - 8 | 1195 | 59 | |
| GenBank mRNA | AB086655.1 | - | 7 - 8 | 1174 | 59 | |
| AF477481.1 | - | 7 - 8 | 1195 | 59 | ||
| AK012041.1 | - | 7 - 8 | 1172 | 59 | ||
| AK017609.1 | - | 7 - 8 | 1169 | 59 | ||
| AK028287.1 | - | 7 - 8 | 1167 | 59 | ||
| AK049163.1 | - | 7 - 8 | 1167 | 59 | ||
| AK076527.1 | - | 6 - 7 | 1277 | 59 | ||
| AK083015.1 | - | 7 - 8 | 1170 | 59 | ||
| BC013842.1 | - | 7 - 8 | 1150 | 59 | ||
| BC024485.1 | - | 4 - 5 | 446 | 59 | ||
| BC048076.1 | - | 7 - 8 | 1195 | 6307476 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdt1
Genomic Map