Assay Details
| Gene Symbol: Foxp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110824.1 | NP_001104294.1 | 9 - 10 | 1530 | 59 | |
| NM_001110825.1 | NP_001104295.1 | 9 - 10 | 1530 | 59 | ||
| NM_028767.2 | NP_083043.2 | 9 - 10 | 1527 | 59 | ||
| GenBank mRNA | AB052766.1 | - | 9 - 10 | 1361 | 59 | |
| AK004693.1 | - | 9 - 10 | 1338 | 59 | ||
| AK143194.1 | - | 9 - 10 | 1377 | 59 | ||
| AY135029.1 | - | 9 - 10 | 1318 | 59 | ||
| BC043702.1 | - | 9 - 10 | 1304 | 5351757 | 59 | |
| BC052407.1 | - | 9 - 10 | 1333 | 5716170 | 59 | |
| BC057110.1 | - | 9 - 10 | 1527 | 6417088 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxp4
Genomic Map