Assay Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033398.2 | NP_203971.2 | 3 - 4 | 1059 | 89 | |
| GenBank mRNA | AF304118.1 | - | 3 - 4 | 992 | 89 | |
| AK185118.1 | - | 1 - 2 | 79 | 89 | ||
| BC006067.1 | - | 3 - 4 | 868 | 89 | ||
| BC011503.1 | - | 3 - 4 | 891 | 89 | ||
| BC056629.1 | - | 3 - 4 | 1059 | 5033487 | 89 | |
| BC117737.1 | - | 2 - 3 | 700 | 89 | ||
| EF527404.1 | - | 3 - 4 | 1059 | 89 | ||
| EF527405.1 | - | 3 - 4 | 845 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jmjd6
Genomic Map