Assay Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080457.3 | NP_536705.3 | 7 - 8 | 8205 | 62 | |
| GenBank mRNA | AF218265.1 | - | 7 - 8 | 1510 | 62 | |
| AF441786.1 | - | 7735 | 62 | |||
| BP765127.1 | - | 2 - 3 | 266 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Muc4
Genomic Map