Assay Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080457.3 | NP_536705.3 | 13 - 14 | 9002 | 67 | |
| GenBank mRNA | AF218265.1 | - | 13 - 14 | 2307 | 67 | |
| AF441786.1 | - | 8532 | 67 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Muc4
Genomic Map