Assay Details
| Gene Symbol: Sult1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133670.1 | NP_598431.1 | 2 - 3 | 195 | 95 | |
| GenBank mRNA | AK002700.1 | - | 2 - 3 | 218 | 95 | |
| BC005413.1 | - | 2 - 3 | 195 | 3488459 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sult1a1
Genomic Map