Assay Details
| Gene Symbol: Rrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145620.4 | NP_663595.1 | 2 - 3 | 209 | 77 | |
| GenBank mRNA | AF368232.1 | - | 2 - 3 | 192 | 77 | |
| AK076113.1 | - | 2 - 3 | 211 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rrp9
Genomic Map