Assay Details
| Gene Symbol: Sec14l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146013.1 | NP_666125.1 | 3 - 4 | 242 | 80 | |
| GenBank mRNA | AK028954.1 | - | 3 - 4 | 203 | 80 | |
| BC026948.1 | - | 3 - 4 | 242 | 5136664 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l4
Genomic Map