Assay Details
| Gene Symbol: Neil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146208.2 | NP_666320.1 | 3 - 4 | 563 | 80 | |
| GenBank mRNA | AB072931.1 | - | 3 - 4 | 430 | 80 | |
| AK031027.1 | - | 3 - 4 | 464 | 80 | ||
| BC024921.1 | - | 3 - 4 | 422 | 3492134 | 80 | |
| BC034753.1 | - | 3 - 4 | 506 | 4945750 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Neil3
Genomic Map