Assay Details
| Gene Symbol: Neil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146208.2 | NP_666320.1 | 7 - 8 | 1196 | 85 | |
| GenBank mRNA | AB072931.1 | - | 7 - 8 | 1063 | 85 | |
| BC024921.1 | - | 7 - 8 | 1055 | 3492134 | 85 | |
| BC034753.1 | - | 7 - 8 | 1139 | 4945750 | 85 | |
| DV646606.1 | - | 5 - 6 | 643 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Neil3
Genomic Map