Assay Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170593.3 | NP_733481.1 | 2 - 3 | 555 | 58 | |
| GenBank mRNA | AK036480.1 | - | 536 | 58 | ||
| AK038938.1 | - | 2 - 3 | 518 | 58 | ||
| AK045560.1 | - | 556 | 58 | |||
| AY150699.1 | - | 2 - 3 | 356 | 58 | ||
| BC118526.1 | - | 2 - 3 | 350 | 40099214 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Disp2
Genomic Map