Assay Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170593.3 | NP_733481.1 | 5 - 6 | 827 | 114 | |
| GenBank mRNA | AK035975.1 | - | 2 - 3 | 1516 | 114 | |
| AK036480.1 | - | 808 | 114 | |||
| AK038938.1 | - | 5 - 6 | 791 | 114 | ||
| AK045560.1 | - | 828 | 114 | |||
| AY150699.1 | - | 5 - 6 | 628 | 114 | ||
| BC118526.1 | - | 5 - 6 | 622 | 40099214 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Disp2
Genomic Map