Assay Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170593.3 | NP_733481.1 | 6 - 7 | 952 | 80 | |
| GenBank mRNA | AK035975.1 | - | 3 - 4 | 1641 | 80 | |
| AK036480.1 | - | 933 | 80 | |||
| AK038938.1 | - | 6 - 7 | 916 | 80 | ||
| AK045560.1 | - | 953 | 80 | |||
| AY150699.1 | - | 6 - 7 | 753 | 80 | ||
| BC118526.1 | - | 6 - 7 | 747 | 40099214 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Disp2
Genomic Map