Assay Details
| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170778.2 | NP_740748.1 | 9 - 10 | 1063 | 64 | |
| GenBank mRNA | BC039699.1 | - | 9 - 10 | 1064 | 4235971 | 64 |
| BC042543.1 | - | 5 - 6 | 528 | 64 | ||
More Information
Set Membership: |
|

Dpyd
Genomic Map