Assay Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136062.2 | NP_001129534.1 | 2 - 3 | 179 | 79 | |
| NM_001276285.1 | NP_001263214.1 | 2 - 3 | 191 | 79 | ||
| NM_007933.3 | NP_031959.1 | 2 - 3 | 291 | 79 | ||
| GenBank mRNA | AK002485.1 | - | 2 - 3 | 151 | 79 | |
| BC013460.1 | - | 1 - 2 | 163 | 4217678 | 79 | |
| BY115250.1 | - | 2 - 3 | 164 | 79 | ||
| BY151264.1 | - | 2 - 3 | 264 | 79 | ||
| CT010193.1 | - | 1 - 2 | 85 | 79 | ||
| X57747.1 | - | 2 - 3 | 142 | 79 | ||
| X62667.1 | - | 1 - 2 | 135 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno3
Genomic Map