Assay Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136062.2 | NP_001129534.1 | 6 - 7 | 540 | 54 | |
| NM_001276285.1 | NP_001263214.1 | 6 - 7 | 552 | 54 | ||
| NM_007933.3 | NP_031959.1 | 6 - 7 | 652 | 54 | ||
| GenBank mRNA | AK002485.1 | - | 6 - 7 | 512 | 54 | |
| BC013460.1 | - | 5 - 6 | 524 | 4217678 | 54 | |
| M20745.1 | - | 3 - 4 | 272 | 54 | ||
| X57747.1 | - | 6 - 7 | 503 | 54 | ||
| X62667.1 | - | 5 - 6 | 496 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977533 | X57747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977533 | X62667.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977533 | M20745.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977533 | NM_007933.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977533 | BC013460.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76018 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977533 | X57747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977533 | X62667.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977533 | M20745.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977533 | NM_007933.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977533 | BC013460.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328539 | X57747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328539 | X62667.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328539 | M20745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328539 | NM_007933.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328539 | BC013460.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27901933 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07389188 | - | - |
| Whole Mouse Genome G4122A | A_51_P328539 | X57747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P328539 | X62667.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P328539 | M20745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P328539 | NM_007933.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P328539 | BC013460.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09367745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27901979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09367763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09080448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09367746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27901962 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10377949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10377942 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417951_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377938 | - | - |
| Mouse Genome 430 2.0 Array | 1449631_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417951_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449631_at | - | - |
| Mouse Genome 430 2.0 Array | 1417951_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13808_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13808_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100008617.mm.1 | NM_001136062.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008617.mm.1 | NM_001276285.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008617.mm.1 | NM_007933.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100008617.mm.1 | TC1100000964.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417951_PM_at | - | - |
| Mouse Expression Set 430 | 1417951_at | - | - |
| Murine Genome U74v2 Set | 96344_PM_at | - | - |
| Murine Genome U74v2 Set | 96344_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417951_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417951_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417951_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417951_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5270136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5270136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005270136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2757569 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2757569 | - | - |
| mouse-6 v1.1 (Illumina) | scl013808.8_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5270136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013808.8_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5270136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2757569 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757569 | - | - |

Eno3
Genomic Map