Assay Details
| Gene Symbol: Mpzl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007962.4 | NP_031988.2 | 2 - 3 | 613 | 80 | |
| GenBank mRNA | AF030454.1 | - | 2 - 3 | 394 | 80 | |
| AK032788.1 | - | 2 - 3 | 394 | 80 | ||
| AK166341.1 | - | 2 - 3 | 344 | 80 | ||
| AK167263.1 | - | 2 - 3 | 377 | 80 | ||
| AK170816.1 | - | 2 - 3 | 379 | 80 | ||
| BC015076.1 | - | 2 - 3 | 439 | 4188907 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpzl2
Genomic Map