Assay Details
| Gene Symbol: Hemt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ011022.1 | - | 1 - 2 | 286 | 72 | |
| BC119337.1 | - | 1 - 2 | 279 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hemt1
Genomic Map