Assay Details
| Gene Symbol: Hey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010423.2 | NP_034553.2 | 3 - 4 | 413 | 93 | |
| GenBank mRNA | AB041590.1 | - | 3 - 4 | 306 | 93 | |
| AF151521.1 | - | 3 - 4 | 361 | 93 | ||
| AF172286.1 | - | 3 - 4 | 394 | 93 | ||
| AF176423.1 | - | 3 - 4 | 375 | 93 | ||
| AF232241.1 | - | 3 - 4 | 393 | 93 | ||
| AJ243895.1 | - | 3 - 4 | 361 | 93 | ||
| AK011149.1 | - | 3 - 4 | 415 | 93 | ||
| AK140212.1 | - | 3 - 4 | 414 | 93 | ||
| BC086635.1 | - | 3 - 4 | 399 | 6809680 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hey1
Genomic Map