Assay Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077705.2 | NP_001071173.1 | 4 - 5 | 715 | 63 | |
| NM_013545.3 | NP_038573.2 | 4 - 5 | 713 | 63 | ||
| GenBank mRNA | AK132509.1 | - | 4 - 5 | 632 | 63 | |
| AK150509.1 | - | 4 - 5 | 620 | 63 | ||
| AK151077.1 | - | 4 - 5 | 634 | 63 | ||
| AK151623.1 | - | 4 - 5 | 637 | 63 | ||
| AK159686.1 | - | 4 - 5 | 672 | 63 | ||
| AK159833.1 | - | 4 - 5 | 665 | 63 | ||
| AK169804.1 | - | 4 - 5 | 715 | 63 | ||
| BC012660.1 | - | 4 - 5 | 658 | 4010462 | 63 | |
| M68902.1 | - | 4 - 5 | 665 | 63 | ||
| M90389.1 | - | 4 - 5 | 615 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn6
Genomic Map