Assay Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016876.3 | NP_058572.2 | 5 - 6 | 330 | 67 | |
| GenBank mRNA | AK038249.1 | - | 5 - 6 | 340 | 67 | |
| AK168362.1 | - | 5 - 6 | 333 | 67 | ||
| U70733.1 | - | 5 - 6 | 320 | 67 | ||
More Information
Set Membership: |
|

Eif3g
Genomic Map