Assay Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016876.3 | NP_058572.2 | 8 - 9 | 729 | 112 | |
| GenBank mRNA | AF108214.1 | - | 8 - 9 | 743 | 112 | |
| AK038249.1 | - | 8 - 9 | 739 | 112 | ||
| BC008511.1 | - | 8 - 9 | 729 | 3592537 | 112 | |
| U70733.1 | - | 8 - 9 | 719 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344425 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | M88758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | U70733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | M88758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P489229 | U70733.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799444 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | M88758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | U70733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | M88758.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P489229 | U70733.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344418 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06717223 | - | - |
| Whole Mouse Genome G4122A | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | M88758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | U70733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | AK038249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | BC008511.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | AK168362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | M88758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P489229 | U70733.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140915 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07621222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07883451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07676962 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07676961 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08197419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07535451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07505364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07578703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07875891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07505368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07578711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07930936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07883451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07930934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07578715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07875890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07578704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344411 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591358 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591359 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417718_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591357 | - | - |
| Mouse Genome 430 2.0 Array | 1417718_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417718_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53356_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53356_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015493.mm.1 | NM_016876.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015493.mm.1 | NM_016876.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015493.mm.1 | TC0900001850.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900015494.mm.1 | NM_016876.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015494.mm.1 | NM_016876.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015494.mm.1 | TC0900001850.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900015492.mm.1 | NM_016876.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015492.mm.1 | NM_016876.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015492.mm.1 | TC0900001850.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417718_PM_at | - | - |
| Mouse Expression Set 430 | 1417718_at | - | - |
| Murine Genome U74v2 Set | 96883_PM_at | - | - |
| Murine Genome U74v2 Set | 96883_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417718_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417718_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417718_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417718_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 593628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3840017 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6400519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6400519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3840017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632985 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2632985 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737193 | - | - |
| mouse-6 v1.1 (Illumina) | scl053356.4_13-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0027558.1_128-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl053356.4_13-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0027558.1_128-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400519 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840017 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737192 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737193 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737192 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737193 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0027558.1_128-S | - | - |

Eif3g
Genomic Map