Assay Details
| Gene Symbol: Ear4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017422.1 | NP_001017422.1 | 1 | 350 | 136 | |
| Gene Symbol: Ear14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017389.2 | NP_059085.2 | 1 | 344 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087193 | NM_017389.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087193 | NM_001017422.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156919 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156919 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087193 | NM_017389.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087193 | NM_001017422.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600616 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156919 | - | - |
| Whole Mouse Genome G4122A | A_51_P305681 | NM_001017422.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109401 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11207750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11207744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11224263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11207749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29600594 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414554 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450615_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414553 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414553 | - | - |
| Mouse Genome 430 2.0 Array | 1450615_at | - | - |
| Mouse Genome 430 2.0 Array | 1450615_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450615_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450615_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 503847_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 503847_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400004555.mm.1 | NM_017389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004555.mm.1 | NM_017389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004555.mm.1 | TC1400000583.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450615_at | - | - |
| Mouse Expression Set 430 | 1450615_PM_at | - | - |
| Murine Genome U74v2 Set | 101715_at | - | - |
| Murine Genome U74v2 Set | 101715_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450615_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450615_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450615_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450615_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493175 | - | - |
| ABI Mouse Genome Survey Microarray | 684705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3290563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773201 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773201 | - | - |
| mouse-6 v1.1 (Illumina) | scl2745.1.1_216-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl2745.1.1_216-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2773201 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2868480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2868480 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773201 | - | - |

Ear4
Genomic Map