Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 1 - 2 | 156 | 113 | |
| GenBank mRNA | AB009294.1 | - | 1 - 2 | 156 | 113 | |
| AF099017.1 | - | 1 - 2 | 116 | 113 | ||
| AF224724.1 | - | 1 - 2 | 116 | 113 | ||
| AK007987.1 | - | 1 - 2 | 154 | 113 | ||
| AK144554.1 | - | 1 - 2 | 128 | 113 | ||
| AK149522.1 | - | 1 - 2 | 120 | 113 | ||
| AK165433.1 | - | 1 - 2 | 133 | 113 | ||
| AK169989.1 | - | 1 - 2 | 152 | 113 | ||
| BC019376.1 | - | 1 - 2 | 121 | 5134851 | 113 | |
| BY202513.1 | - | 1 - 2 | 199 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map