Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 2 - 3 | 337 | 104 | |
| GenBank mRNA | AB009294.1 | - | 2 - 3 | 337 | 104 | |
| AF099017.1 | - | 2 - 3 | 297 | 104 | ||
| AF224724.1 | - | 2 - 3 | 297 | 104 | ||
| AK007987.1 | - | 2 - 3 | 335 | 104 | ||
| AK144554.1 | - | 2 - 3 | 309 | 104 | ||
| AK149522.1 | - | 2 - 3 | 301 | 104 | ||
| AK165433.1 | - | 2 - 3 | 314 | 104 | ||
| AK169989.1 | - | 2 - 3 | 333 | 104 | ||
| BC019376.1 | - | 2 - 3 | 302 | 5134851 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map