Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 6 - 7 | 764 | 64 | |
| GenBank mRNA | AF099017.1 | - | 6 - 7 | 724 | 64 | |
| AF224724.1 | - | 6 - 7 | 724 | 64 | ||
| AK007987.1 | - | 6 - 7 | 762 | 64 | ||
| AK075718.1 | - | 6 - 7 | 673 | 64 | ||
| AK144554.1 | - | 6 - 7 | 736 | 64 | ||
| AK149522.1 | - | 6 - 7 | 728 | 64 | ||
| AK169989.1 | - | 6 - 7 | 760 | 64 | ||
| BC019376.1 | - | 6 - 7 | 729 | 5134851 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map