Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 7 - 8 | 879 | 62 | |
| GenBank mRNA | AF099017.1 | - | 7 - 8 | 839 | 62 | |
| AF224724.1 | - | 7 - 8 | 839 | 62 | ||
| AK007987.1 | - | 7 - 8 | 877 | 62 | ||
| AK075718.1 | - | 7 - 8 | 788 | 62 | ||
| AK144554.1 | - | 7 - 8 | 850 | 62 | ||
| AK149522.1 | - | 7 - 8 | 843 | 62 | ||
| AK165433.1 | - | 5 - 6 | 1189 | 62 | ||
| AK169989.1 | - | 7 - 8 | 875 | 62 | ||
| BC019376.1 | - | 7 - 8 | 844 | 5134851 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map