Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 8 - 9 | 1056 | 93 | |
| GenBank mRNA | AF099017.1 | - | 8 - 9 | 1016 | 93 | |
| AF224724.1 | - | 8 - 9 | 1016 | 93 | ||
| AK007987.1 | - | 8 - 9 | 1054 | 93 | ||
| AK075718.1 | - | 8 - 9 | 965 | 93 | ||
| AK144554.1 | - | 8 - 9 | 1027 | 93 | ||
| AK149522.1 | - | 8 - 9 | 1020 | 93 | ||
| AK165433.1 | - | 6 - 7 | 1366 | 93 | ||
| AK169989.1 | - | 8 - 9 | 1052 | 93 | ||
| BC019376.1 | - | 8 - 9 | 1021 | 5134851 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map