Assay Details
| Gene Symbol: Hhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020259.4 | NP_064655.4 | 8 - 9 | 1918 | 88 | |
| GenBank mRNA | AF116865.1 | - | 8 - 9 | 1899 | 88 | |
| AK031539.1 | - | 8 - 9 | 1915 | 88 | ||
| AK031841.1 | - | 8 - 9 | 1428 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hhip
Genomic Map