Assay Details
| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023158.6 | NP_075647.3 | 4 - 5 | 1237 | 70 | |
| GenBank mRNA | AF277001.1 | - | 4 - 5 | 700 | 70 | |
| AF301017.1 | - | 4 - 5 | 766 | 70 | ||
| AK003085.1 | - | 4 - 5 | 896 | 70 | ||
| AK009599.1 | - | 4 - 5 | 786 | 70 | ||
| AK028875.1 | - | 4 - 5 | 1239 | 70 | ||
| AK030515.1 | - | 4 - 5 | 662 | 70 | ||
| AK151330.1 | - | 4 - 5 | 1011 | 70 | ||
| AK152102.1 | - | 4 - 5 | 1011 | 70 | ||
| AK152547.1 | - | 4 - 5 | 1189 | 70 | ||
| AK153192.1 | - | 4 - 5 | 953 | 70 | ||
| AK170428.1 | - | 5 - 6 | 1408 | 70 | ||
| BC019961.1 | - | 4 - 5 | 1118 | 3988542 | 70 | |
| GQ380016.1 | - | 5 - 6 | 803 | 70 | ||
| GQ380017.1 | - | 6 - 7 | 854 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
receptor-mediated endocytosis
chemotaxis
T cell chemotaxis
positive regulation of cell growth
positive regulation of cell migration
response to cytokine
response to interferon-gamma
response to tumor necrosis factor
lymphocyte chemotaxis
cell chemotaxis
cellular response to lipopolysaccharide
low-density lipoprotein receptor activity
scavenger receptor activity
cytokine activity
chemokine activity
chemokine receptor binding
receptor-mediated endocytosis
chemotaxis
T cell chemotaxis
positive regulation of cell growth
positive regulation of cell migration
response to cytokine
response to interferon-gamma
response to tumor necrosis factor
lymphocyte chemotaxis
cell chemotaxis
cellular response to lipopolysaccharide
low-density lipoprotein receptor activity
scavenger receptor activity
cytokine activity
chemokine activity
chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900883 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186716 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105809 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374203 | BC019961.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374203 | GQ380017.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374203 | AK152102.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374203 | AK152547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P374203 | AK153192.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900872 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105809 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07388585 | - | - |
| Whole Mouse Genome G4122A | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P99888 | AK028875.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P99888 | NM_023158.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374203 | BC019961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374203 | GQ380017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374203 | AK152102.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374203 | AK152547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P374203 | AK153192.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114820 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418250 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418248 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09418238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09340654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09418254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09596430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09483950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27900881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09112640 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387892 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418718_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449195_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387890 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10377826 | - | - |
| Mouse Genome 430 2.0 Array | 1418718_at | - | - |
| Mouse Genome 430 2.0 Array | 1418718_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449195_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1449195_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66102_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66102_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100028610.mm.1 | NM_023158.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028610.mm.1 | NM_023158.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028610.mm.1 | TC1100003097.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028606.mm.1 | NM_023158.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028606.mm.1 | NM_023158.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028606.mm.1 | TC1100003097.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028608.mm.1 | NM_023158.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028608.mm.1 | NM_023158.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028608.mm.1 | TC1100003097.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028605.mm.1 | NM_023158.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028605.mm.1 | NM_023158.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028605.mm.1 | TC1100003097.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028607.mm.1 | NM_023158.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028607.mm.1 | NM_023158.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028607.mm.1 | TC1100003097.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418718_PM_at | - | - |
| Mouse Expression Set 430 | 1418718_at | - | - |
| Mouse Expression Set 430 | 1449195_PM_s_at | - | - |
| Mouse Expression Set 430 | 1449195_s_at | - | - |
| Murine Genome U74v2 Set | 134771_at | - | - |
| Murine Genome U74v2 Set | 134771_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449195_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449195_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418718_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418718_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449195_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418718_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449195_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418718_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101780301 | - | - |
| MouseWG-6 v2 Expression BeadChip | 510278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100450458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101780301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687586 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2687586 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254723 | - | - |
| mouse-6 v1.1 (Illumina) | scl41338.15.1_31-S | - | - |
| mouse-6 v1.1 (Illumina) | scl39995.5_256-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0066102.2_212-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510278 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101780301 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100450458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066102.2_212-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101780301 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100450458 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2687586 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2687586 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2461299 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254723 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066102.2_212-S | - | - |

Cxcl16
Genomic Map