Assay Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168333.1 | NP_001161805.1 | 4 - 5 | 554 | 77 | |
| NM_023476.3 | NP_075965.2 | 4 - 5 | 695 | 77 | ||
| GenBank mRNA | AB047402.1 | - | 4 - 5 | 696 | 77 | |
| AB050626.1 | - | 4 - 5 | 532 | 77 | ||
| AK087142.1 | - | 554 | 77 | |||
| BC018539.1 | - | 4 - 5 | 539 | 3596560 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tinagl1
Genomic Map